Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMT1;3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113336.3 | NP_189073.1 | 1 | 1599 | 133 | |
GenBank mRNA | AK227798.1 | - | 1 | 1527 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |