Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113489.3 | NP_189215.1 | 1 - 2 | 1106 | 85 | |
GenBank mRNA | AF066079.1 | - | 1 - 2 | 965 | 85 | |
AY037262.1 | - | 1 - 2 | 979 | 85 | ||
BT002343.1 | - | 1 - 2 | 907 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |