Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126027.1 | NP_001119499.1 | 5 - 6 | 549 | 91 | |
NM_001203688.1 | NP_001190617.1 | 5 - 6 | 569 | 91 | ||
NM_001345665.1 | NP_001332256.1 | 5 - 6 | 549 | 91 | ||
NM_001345666.1 | NP_001332257.1 | 3 - 4 | 360 | 91 | ||
NM_125906.4 | NP_201312.2 | 4 - 5 | 523 | 91 | ||
GenBank mRNA | AY231450.1 | - | 4 - 5 | 456 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |