Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DET3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101154.4 | NP_563916.1 | 8 - 9 | 2244 | 70 | |
GenBank mRNA | AB493453.1 | - | 8 - 9 | 865 | 70 | |
AF208261.1 | - | 8 - 9 | 874 | 70 | ||
AF334725.1 | - | 8 - 9 | 865 | 70 | ||
AY059872.1 | - | 8 - 9 | 932 | 70 | ||
AY093334.1 | - | 8 - 9 | 865 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |