Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101201.2 | NP_563926.1 | 3 - 4 | 721 | 147 | |
GenBank mRNA | AF367336.1 | - | 3 - 4 | 709 | 147 | |
AY124865.1 | - | 3 - 4 | 622 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |