Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ELF5A-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101261.3 | NP_172848.1 | 2 - 3 | 438 | 148 | |
GenBank mRNA | AF296082.1 | - | 2 - 3 | 296 | 148 | |
AY063780.1 | - | 2 - 3 | 281 | 148 | ||
AY117272.1 | - | 2 - 3 | 241 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |