Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106129.4 | NP_565090.1 | 4 - 5 | 1042 | 98 | |
NM_202414.1 | NP_974143.1 | 4 - 5 | 1041 | 98 | ||
GenBank mRNA | AF367342.1 | - | 4 - 5 | 1040 | 98 | |
AY056055.1 | - | 4 - 5 | 1042 | 98 | ||
AY124864.1 | - | 4 - 5 | 946 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |