Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084367.2 | NP_001077836.1 | 7 - 8 | 3303 | 87 | |
NM_106379.9 | NP_177854.6 | 7 - 8 | 3455 | 87 | ||
GenBank mRNA | EU014690.1 | - | 6 - 7 | 3168 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |