Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334784.1 | NP_001321064.1 | 3 - 4 | 581 | 70 | |
NM_106398.3 | NP_177873.1 | 3 - 4 | 623 | 70 | ||
GenBank mRNA | AK229693.1 | - | 3 - 4 | 499 | 70 | |
AY085554.1 | - | 3 - 4 | 513 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |