Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334818.1 | NP_001322316.1 | 1 - 2 | 353 | 110 | |
NM_106458.4 | NP_177932.1 | 1 - 2 | 439 | 110 | ||
GenBank mRNA | BT002801.1 | - | 1 - 2 | 348 | 110 | |
BT004348.1 | - | 1 - 2 | 287 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |