Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198504.2 | NP_001185433.1 | 2 - 3 | 613 | 81 | |
NM_001334866.1 | NP_001322202.1 | 2 - 3 | 561 | 81 | ||
NM_106550.4 | NP_565197.3 | 2 - 3 | 729 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |