Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH5F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106592.5 | NP_178062.1 | 16 - 17 | 1420 | 100 | |
GenBank mRNA | AF117335.1 | - | 16 - 17 | 1338 | 100 | |
AF428367.1 | - | 16 - 17 | 1340 | 100 | ||
AY056147.1 | - | 16 - 17 | 1326 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |