Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATERDJ3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337788.1 | NP_001319504.1 | 5 - 6 | 1185 | 105 | |
NM_111731.4 | NP_187509.1 | 5 - 6 | 1211 | 105 | ||
GenBank mRNA | BT003887.1 | - | 5 - 6 | 1127 | 105 | |
BT005001.1 | - | 5 - 6 | 1075 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |