Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GATL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113753.5 | NP_189474.2 | 1 | 1220 | 67 | |
GenBank mRNA | AK176597.1 | - | 1 | 1087 | 67 | |
BT011750.1 | - | 1 | 1028 | 67 | ||
KJ138808.1 | - | 1 | 1028 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |