Search Thermo Fisher Scientific
Gene Symbol: PHOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035737.2 | NP_001030814.1 | 5 - 6 | 1668 | 106 | |
NM_114447.4 | NP_190164.1 | 6 - 7 | 1666 | 106 | ||
GenBank mRNA | AF030864.1 | - | 6 - 7 | 1521 | 106 | |
AF360218.1 | - | 5 - 6 | 1475 | 106 | ||
AY040062.1 | - | 5 - 6 | 1458 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |