Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339367.1 | NP_001326126.1 | 3 - 4 | 1421 | 104 | |
NM_114697.4 | NP_566902.1 | 4 - 5 | 1219 | 104 | ||
GenBank mRNA | AK226596.1 | - | 4 - 5 | 1007 | 104 | |
AY040015.1 | - | 4 - 5 | 1005 | 104 | ||
AY079408.1 | - | 4 - 5 | 952 | 104 | ||
AY316675.1 | - | 4 - 5 | 952 | 104 | ||
BT000678.1 | - | 4 - 5 | 998 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |