Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122181.3 | NP_197667.1 | 15 - 16 | 2739 | 89 | |
GenBank mRNA | AK228695.1 | - | 15 - 16 | 2713 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |