Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CaS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343788.1 | NP_001331124.1 | 2 - 3 | 586 | 69 | |
NM_122212.5 | NP_197697.1 | 2 - 3 | 586 | 69 | ||
GenBank mRNA | AY045798.1 | - | 2 - 3 | 561 | 69 | |
AY079331.1 | - | 2 - 3 | 463 | 69 | ||
AY341888.1 | - | 2 - 3 | 548 | 69 | ||
BT000666.1 | - | 2 - 3 | 565 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |