Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203448.2 | NP_001190377.1 | 3 - 4 | 532 | 83 | |
NM_001343862.1 | NP_001331303.1 | 2 - 3 | 3349 | 83 | ||
NM_001343863.1 | NP_001331302.1 | 1 - 2 | 815 | 83 | ||
NM_122333.6 | NP_197815.1 | 3 - 4 | 600 | 83 | ||
GenBank mRNA | AF192886.1 | - | 2 - 3 | 171 | 83 | |
AY063993.1 | - | 3 - 4 | 261 | 83 | ||
AY096693.1 | - | 2 - 3 | 171 | 83 | ||
Y18870.1 | - | 284 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |