Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102057.4 | NP_564156.1 | 7 - 8 | 1287 | 56 | |
GenBank mRNA | BT026034.1 | - | 6 - 7 | 769 | 56 | |
L10209.1 | - | 7 - 8 | 850 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |