Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATSAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102109.4 | NP_173676.2 | 2 - 3 | 535 | 98 | |
GenBank mRNA | AF266460.1 | - | 2 - 3 | 333 | 98 | |
AY227244.1 | - | 2 - 3 | 333 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |