Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332600.1 | NP_001320962.1 | 4 - 5 | 560 | 77 | |
NM_102191.6 | NP_564194.1 | 4 - 5 | 519 | 77 | ||
GenBank mRNA | AB493475.1 | - | 4 - 5 | 434 | 77 | |
AF195047.1 | - | 4 - 5 | 461 | 77 | ||
BT026410.1 | - | 4 - 5 | 434 | 77 | ||
DQ446281.1 | - | 4 - 5 | 434 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |