Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCH;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344020.1 | NP_001330468.1 | 11 - 12 | 1468 | 91 | |
NM_122644.5 | NP_198114.2 | 12 - 13 | 1399 | 91 | ||
GenBank mRNA | AB051072.1 | - | 12 - 13 | 924 | 91 | |
AK118707.1 | - | 12 - 13 | 1381 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |