Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122961.4 | NP_198420.1 | 9 - 10 | 1326 | 124 | |
GenBank mRNA | AY045780.1 | - | 9 - 10 | 1304 | 124 | |
AY142570.1 | - | 8 - 9 | 1101 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |