Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAUT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124152.3 | NP_568688.1 | 4 - 5 | 371 | 65 | |
GenBank mRNA | AY056202.1 | - | 4 - 5 | 319 | 65 | |
AY091394.1 | - | 4 - 5 | 254 | 65 | ||
KJ138647.1 | - | 4 - 5 | 254 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |