Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: BIK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129522.5 | NP_181496.1 | 4 - 5 | 1052 | 96 | |
GenBank mRNA | AF325086.1 | - | 4 - 5 | 636 | 96 | |
AY062493.1 | - | 4 - 5 | 854 | 96 | ||
AY065029.1 | - | 4 - 5 | 854 | 96 | ||
AY093278.1 | - | 4 - 5 | 636 | 96 | ||
AY093997.1 | - | 4 - 5 | 636 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |