Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPB5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336901.1 | NP_001324726.1 | 1 - 2 | 538 | 89 | |
NM_001336902.1 | NP_001324725.1 | 1 - 2 | 511 | 89 | ||
NM_129697.3 | NP_181665.1 | 1 - 2 | 511 | 89 | ||
GenBank mRNA | AY078926.1 | - | 1 - 2 | 263 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |