Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP47C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103642.3 | NP_175180.1 | 1 - 2 | 707 | 119 | |
NM_202255.2 | NP_973984.1 | 1 - 2 | 707 | 119 | ||
GenBank mRNA | AF344325.1 | - | 1 - 2 | 379 | 119 | |
AY035180.1 | - | 1 - 2 | 511 | 119 | ||
AY062960.1 | - | 1 - 2 | 379 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |