Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP47C' | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333273.1 | NP_001321088.1 | 1 - 2 | 565 | 137 | |
NM_103643.3 | NP_175181.1 | 1 - 2 | 632 | 137 | ||
GenBank mRNA | AK227406.1 | - | 1 - 2 | 496 | 137 | |
BT030029.1 | - | 1 - 2 | 383 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |