Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332792.1 | NP_001322427.1 | 1 - 2 | 379 | 151 | |
NM_102559.2 | NP_174115.1 | 1 - 2 | 379 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |