Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SARNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042654.2 | NP_001036119.1 | 10 - 11 | 602 | 74 | |
GenBank mRNA | BC109532.1 | - | 10 - 11 | 603 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |