Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001157.1 | NP_001001157.1 | 2 - 3 | 1115 | 65 | |
GenBank mRNA | AB119637.1 | - | 2 - 3 | 1115 | 65 | |
AJ491863.1 | - | 1 - 2 | 107 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Bos taurus Inventoried Probe spans exons |