Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GYPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002886.1 | NP_001002886.1 | 2 - 3 | 211 | 78 | |
GenBank mRNA | AY234181.1 | - | 2 - 3 | 211 | 78 | |
BC114794.1 | - | 2 - 3 | 181 | 78 | ||
DQ324372.1 | - | 2 - 3 | 207 | 78 | ||
DQ324373.1 | - | 2 - 3 | 207 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |