Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: SPATA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011677.1 | NP_001011677.1 | 2 - 3 | 433 | 147 | |
GenBank mRNA | AY841904.1 | - | 2 - 3 | 433 | 147 | |
BC102855.1 | - | 2 - 3 | 382 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |