Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LRRC51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024545.1 | NP_001019716.1 | 2 - 3 | 161 | 69 | |
NM_001282038.1 | NP_001268967.1 | 2 - 3 | 122 | 69 | ||
GenBank mRNA | BT020631.1 | - | 2 - 3 | 161 | 69 | |
EE348175.1 | - | 2 - 3 | 122 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |