Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174019.2 | NP_776444.1 | 2 - 3 | 993 | 82 | |
GenBank mRNA | GQ179990.1 | - | 2 - 3 | 924 | 82 | |
U08018.1 | - | 2 - 3 | 993 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |