Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174237.2 | NP_776662.1 | 1 - 2 | 284 | 118 | |
GenBank mRNA | BC114157.1 | - | 1 - 2 | 251 | 118 | |
M25757.1 | - | 1 - 2 | 284 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |