Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174249.2 | NP_776674.1 | 2 - 3 | 129 | 60 | |
GenBank mRNA | BC126817.1 | - | 2 - 3 | 141 | 60 | |
X51700.1 | - | 129 | 60 | |||
X53699.1 | - | 129 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Inventoried Probe spans exons |