Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174289.2 | NP_776714.1 | 1 - 2 | 217 | 103 | |
GenBank mRNA | M26142.1 | - | 1 - 2 | 217 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |