Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174671.2 | NP_777096.1 | 6 - 7 | 790 | 60 | |
GenBank mRNA | BC112481.1 | - | 6 - 7 | 58 | ||
BC142283.1 | - | 6 - 7 | 866 | 60 | ||
BC146129.1 | - | 6 - 7 | 805 | 60 | ||
BT020806.1 | - | 6 - 7 | 745 | 60 | ||
BT030473.1 | - | 6 - 7 | 736 | 60 | ||
M58703.1 | - | 6 - 7 | 790 | 60 | ||
M60662.1 | - | 6 - 7 | 790 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |