Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177486.2 | NP_803452.1 | 1 - 2 | 334 | 110 | |
GenBank mRNA | AF022073.1 | - | 1 - 2 | 334 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |