Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: EIF2B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015593.1 | NP_001015593.1 | 4 - 5 | 626 | 68 | |
GenBank mRNA | BC123823.1 | - | 4 - 5 | 630 | 68 | |
BT021006.1 | - | 4 - 5 | 626 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |