Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034587.1 | NP_001029759.1 | 2 - 3 | 1112 | 92 | |
GenBank mRNA | BC123712.1 | - | 2 - 3 | 811 | 92 | |
BT021814.1 | - | 2 - 3 | 1112 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |