Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TOLLIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039961.1 | NP_001035050.1 | 4 - 5 | 583 | 71 | |
GenBank mRNA | BC123457.1 | - | 4 - 5 | 585 | 71 | |
BT021785.1 | - | 4 - 5 | 583 | 71 | ||
DQ319073.1 | - | 4 - 5 | 554 | 71 | ||
DQ407275.1 | - | 4 - 5 | 518 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |