Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MNF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206614.1 | NP_001193543.1 | 2 - 3 | 257 | 65 | |
GenBank mRNA | BC103264.1 | - | 2 - 3 | 233 | 65 | |
CO875952.1 | - | 2 - 3 | 276 | 65 | ||
DT860945.1 | - | 2 - 3 | 257 | 65 | ||
EE221541.1 | - | 2 - 3 | 256 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |