Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078035.2 | NP_001071503.1 | 1 - 2 | 106 | 80 | |
GenBank mRNA | BC102336.1 | - | 1 - 2 | 107 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |