Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GUK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159412.1 | NP_001152884.1 | 6 - 7 | 580 | 72 | |
NM_001159413.1 | NP_001152885.1 | 6 - 7 | 561 | 72 | ||
NM_174078.3 | NP_776503.1 | 6 - 7 | 677 | 72 | ||
GenBank mRNA | BC102478.1 | - | 6 - 7 | 677 | 72 | |
DN283346.1 | - | 6 - 7 | 562 | 72 | ||
X67029.1 | - | 561 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |