Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034282.1 | NP_001029454.1 | 11 - 12 | 1430 | 87 | |
GenBank mRNA | BC103242.1 | - | 11 - 12 | 1430 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |