Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034579.2 | NP_001029751.2 | 6 - 7 | 681 | 150 | |
GenBank mRNA | AM086994.1 | - | 6 - 7 | 681 | 150 | |
BC102770.1 | - | 6 - 7 | 681 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |