Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTCAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035062.2 | NP_001030234.1 | 6 - 7 | 765 | 79 | |
GenBank mRNA | BC103079.1 | - | 6 - 7 | 766 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |